Anti-SLAMF6 Mouse Monoclonal Antibody [clone: 292810]

Supplier: R & D Systems
MAB1908
220019-443EA 594.37 USD
220019-443
Anti-SLAMF6 Mouse Monoclonal Antibody [clone: 292810]
Antibodies
Type: Primary
Antigen: SLAMF6
Clonality: Monoclonal
Clone: 292810
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR