Anti-SLAMF8 Mouse Monoclonal Antibody [clone: 250020]

Supplier: R & D Systems
MAB19073
220019-442EA 772.25 USD
220019-442
Anti-SLAMF8 Mouse Monoclonal Antibody [clone: 250020]
Antibodies
Type: Primary
Antigen: SLAMF8
Clonality: Monoclonal
Clone: 250020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR