Anti-HPRG Rat Monoclonal Antibody [clone: 242425]

Supplier: R & D Systems
MAB1905
220019-439EA 763.57 USD
220019-439
Anti-HPRG Rat Monoclonal Antibody [clone: 242425]
Antibodies
Type: Primary
Antigen: HPRG
Clonality: Monoclonal
Clone: 242425
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR