Anti-CLEC11A Mouse Monoclonal Antibody [clone: 239029]

Supplier: R & D Systems
MAB1904
220019-438EA 763.57 USD
220019-438
Anti-CLEC11A Mouse Monoclonal Antibody [clone: 239029]
Antibodies
Type: Primary
Antigen: CLEC11A
Clonality: Monoclonal
Clone: 239029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR