Anti-KLRF1 Rat Monoclonal Antibody [clone: 239127]

Supplier: R & D Systems
MAB1900
220019-437EA 763.57 USD
220019-437
Anti-KLRF1 Rat Monoclonal Antibody [clone: 239127]
Antibodies
Type: Primary
Antigen: KLRF1
Clonality: Monoclonal
Clone: 239127
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR