Anti-CXCR5 Mouse Monoclonal Antibody [clone: 51505]

Supplier: R & D Systems
MAB190-100
102885-398EA 655.11 USD
102885-398
Anti-CXCR5 Mouse Monoclonal Antibody [clone: 51505]
Antibodies
Type: Primary
Antigen: CXCR5
Clonality: Monoclonal
Clone: 51505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR