Anti-FGFRL1 Rat Monoclonal Antibody [clone: 223003]

Supplier: R & D Systems
MAB1899
220019-435EA 772.25 USD
220019-435
Anti-FGFRL1 Rat Monoclonal Antibody [clone: 223003]
Antibodies
Type: Primary
Antigen Symbol: FGFRL1
Antigen Name: fibroblast growth factor receptor-like 1
Antigen Synonyms: FGFR5, FGFR5beta, FGFR5gamma
Clonality: Monoclonal
Clone: 223003
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR