Anti-BPIFA1 Mouse Monoclonal Antibody [clone: 251512]

Supplier: R & D Systems
MAB1897
220019-433EA 600.15 USD
220019-433
Anti-BPIFA1 Mouse Monoclonal Antibody [clone: 251512]
Antibodies
Type: Primary
Antigen: BPIFA1
Clonality: Monoclonal
Clone: 251512
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR