Anti-TNFSF15 Rat Monoclonal Antibody [clone: 820446]

Supplier: R & D Systems
MAB1896
102885-396EA 594.37 USD
102885-396
Anti-TNFSF15 Rat Monoclonal Antibody [clone: 820446]
Antibodies
Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone: 820446
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR