Anti-APCS Mouse Monoclonal Antibody [clone: 27392]

Supplier: R & D Systems
MAB18951
220019-432EA 763.57 USD
220019-432
Anti-APCS Mouse Monoclonal Antibody [clone: 27392]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: APCS
Clonality: Monoclonal
Clone: 27392
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR