Anti-APCS Mouse Monoclonal Antibody [clone: 273912]

Supplier: R & D Systems
MAB1895
220019-431EA 584.25 USD
220019-431
Anti-APCS Mouse Monoclonal Antibody [clone: 273912]
Antibodies
Type: Primary
Antigen: APCS
Clonality: Monoclonal
Clone: 273912
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR