Anti-PTPN11 Rat Monoclonal Antibody [clone: 255509]

Supplier: R & D Systems
MAB1894
220019-430EA 566.89 USD
220019-430
Anti-PTPN11 Rat Monoclonal Antibody [clone: 255509]
Antibodies
Type: Primary
Antigen: PTPN11
Clonality: Monoclonal
Clone: 255509
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR