Anti-CST7 Mouse Monoclonal Antibody [clone: 292103]

Supplier: R & D Systems
MAB1889
220019-429EA 577.02 USD
220019-429
Anti-CST7 Mouse Monoclonal Antibody [clone: 292103]
Antibodies
Type: Primary
Antigen: CST7
Clonality: Monoclonal
Clone: 292103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR