Anti-SCARB2 Rat Monoclonal Antibody [clone: 220411]

Supplier: R & D Systems
MAB1888
220019-428EA 763.57 USD
220019-428
Anti-SCARB2 Rat Monoclonal Antibody [clone: 220411]
Antibodies
Type: Primary
Antigen: SCARB2
Clonality: Monoclonal
Clone: 220411
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR