Anti-IL23A Rat Monoclonal Antibody [clone: 320234]

Supplier: R & D Systems
MAB1887
220019-427EA 772.25 USD
220019-427
Anti-IL23A Rat Monoclonal Antibody [clone: 320234]
Antibodies
Type: Primary
Antigen: IL23A
Clonality: Monoclonal
Clone: 320234
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR