Anti-CD151 Mouse Monoclonal Antibody [clone: 210127]

Supplier: R & D Systems
MAB1884
220019-425EA 772.25 USD
220019-425
Anti-CD151 Mouse Monoclonal Antibody [clone: 210127]
Antibodies
Type: Primary
Antigen: CD151
Clonality: Monoclonal
Clone: 210127
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR