Anti-PTPN6 Rat Monoclonal Antibody [clone: 255402]

Supplier: R & D Systems
MAB1878
220019-421EA 594.37 USD
220019-421
Anti-PTPN6 Rat Monoclonal Antibody [clone: 255402]
Antibodies
Type: Primary
Antigen: PTPN6
Clonality: Monoclonal
Clone: 255402
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR