Anti-BMP3 Mouse Monoclonal Antibody [clone: 203706]

Supplier: R & D Systems
MAB1876
220019-420EA 772.25 USD
220019-420
Anti-BMP3 Mouse Monoclonal Antibody [clone: 203706]
Antibodies
Type: Primary
Antigen: BMP3
Clonality: Monoclonal
Clone: 203706
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR