Anti-Troponin T type 2 Mouse Monoclonal Antibody [clone: 200805]

Supplier: R & D Systems
MAB1874
220019-418EA 616.06 USD
220019-418
Anti-Troponin T type 2 Mouse Monoclonal Antibody [clone: 200805]
Antibodies
Type: Primary
Antigen: Troponin T type 2
Clonality: Monoclonal
Clone: 200805
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR