Anti-HRG Mouse Monoclonal Antibody [clone: 227901]

Supplier: R & D Systems
MAB1869
220019-417EA 772.25 USD
220019-417
Anti-HRG Mouse Monoclonal Antibody [clone: 227901]
Antibodies
Type: Primary
Antigen: HRG
Clonality: Monoclonal
Clone: 227901
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR