Anti-HDGFL1 Rat Monoclonal Antibody [clone: 249308]

Supplier: R & D Systems
MAB1868
220019-416EA 763.57 USD
220019-416
Anti-HDGFL1 Rat Monoclonal Antibody [clone: 249308]
Antibodies
Type: Primary
Antigen: HDGFL1
Clonality: Monoclonal
Clone: 249308
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR