Anti-GZMB Rat Monoclonal Antibody [clone: 216315]

Supplier: R & D Systems
MAB1865
220019-415EA 763.57 USD
220019-415
Anti-GZMB Rat Monoclonal Antibody [clone: 216315]
Antibodies
Type: Primary
Antigen Symbol: GZMB
Antigen Name: granzyme B
Antigen Synonyms: AI553453, CCP-1/C11, CCP1, Ctla-1, Ctla1, GZB
Clonality: Monoclonal
Clone: 216315
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR