Anti-ITGA5 Mouse Monoclonal Antibody [clone: 612557]

Supplier: R & D Systems
MAB18642
102885-394EA 594.37 USD
102885-394
Anti-ITGA5 Mouse Monoclonal Antibody [clone: 612557]
Antibodies
Type: Primary
Antigen: ITGA5
Clonality: Monoclonal
Clone: 612557
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR