Anti-ITGA5 Mouse Monoclonal Antibody [clone: 238307]

Supplier: R & D Systems
MAB1864
220019-414EA 600.15 USD
220019-414
Anti-ITGA5 Mouse Monoclonal Antibody [clone: 238307]
Antibodies
Adhesion Activation

Type: Primary
Antigen: ITGA5
Clonality: Monoclonal
Clone: 238307
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR