Anti-AMN Mouse Monoclonal Antibody [clone: 2688]

Supplier: R & D Systems
MAB1860
220019-413EA 561.11 USD
220019-413
Anti-AMN Mouse Monoclonal Antibody [clone: 2688]
Antibodies
Type: Primary
Antigen: AMN
Clonality: Monoclonal
Clone: 2688
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR