Anti-CLEC7A Mouse Monoclonal Antibody [clone: 259931]

Supplier: R & D Systems
MAB1859
220019-412EA 655.11 USD
220019-412
Anti-CLEC7A Mouse Monoclonal Antibody [clone: 259931]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: CLEC7A
Clonality: Monoclonal
Clone: 259931
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR