Anti-FASLG Mouse Monoclonal Antibody [clone: 252029]

Supplier: R & D Systems
MAB1858
220019-411EA 763.57 USD
220019-411
Anti-FASLG Mouse Monoclonal Antibody [clone: 252029]
Antibodies
Type: Primary
Antigen Symbol: FASLG
Antigen Name: Fas ligand (TNF superfamily, member 6)
Antigen Synonyms: Apt1Lg1, CD95-L, Fasl, Tnfsf6
Clonality: Monoclonal
Clone: 252029
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR