Anti-CD2 Mouse Monoclonal Antibody [clone: 299812]

Supplier: R & D Systems
MAB18561
220019-409EA 594.37 USD
220019-409
Anti-CD2 Mouse Monoclonal Antibody [clone: 299812]
Antibodies
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: 299812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR