Anti-CD2 Mouse Monoclonal Antibody [clone: 299813]

Supplier: R & D Systems
MAB1856
220019-408EA 594.37 USD
220019-408
Anti-CD2 Mouse Monoclonal Antibody [clone: 299813]
Antibodies
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: 299813
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR