Anti-CD84 Mouse Monoclonal Antibody [clone: 273508]

Supplier: R & D Systems
MAB1855
220019-407EA 594.37 USD
220019-407
Anti-CD84 Mouse Monoclonal Antibody [clone: 273508]
Antibodies
Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: 273508
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR