Anti-KIR Mouse Monoclonal Antibody [clone: 180704]

Supplier: R & D Systems
MAB1848
220019-403EA 763.57 USD
220019-403
Anti-KIR Mouse Monoclonal Antibody [clone: 180704]
Antibodies
Type: Primary
Antigen: KIR
Clonality: Monoclonal
Clone: 180704
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR