Anti-KIR2DS4 Mouse Monoclonal Antibody [clone: 179315]

Supplier: R & D Systems
MAB1847
220019-402EA 772.25 USD
220019-402
Anti-KIR2DS4 Mouse Monoclonal Antibody [clone: 179315]
Antibodies
Type: Primary
Antigen: KIR2DS4
Clonality: Monoclonal
Clone: 179315
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR