Anti-KIRREL2 Mouse Monoclonal Antibody [clone: 252320]

Supplier: R & D Systems
MAB1846
220019-401EA 594.37 USD
220019-401
Anti-KIRREL2 Mouse Monoclonal Antibody [clone: 252320]
Antibodies
Type: Primary
Antigen: KIRREL2
Clonality: Monoclonal
Clone: 252320
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR