Anti-CFC1 Rat Monoclonal Antibody [clone: 221423]

Supplier: R & D Systems
MAB1840
220019-398EA 763.57 USD
220019-398
Anti-CFC1 Rat Monoclonal Antibody [clone: 221423]
Antibodies
Type: Primary
Antigen: CFC1
Clonality: Monoclonal
Clone: 221423
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR