Anti-TP53 Mouse Monoclonal Antibody [clone: 261366]

Supplier: R & D Systems
MAB18391
220019-396EA 594.37 USD
220019-396
Anti-TP53 Mouse Monoclonal Antibody [clone: 261366]
Antibodies
Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: 261366
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR