Anti-CDH1 Mouse Monoclonal Antibody [clone: 180224]

Supplier: R & D Systems
MAB18381
220019-393EA 633.42 USD
220019-393
Anti-CDH1 Mouse Monoclonal Antibody [clone: 180224]
Antibodies
Type: Primary
Antigen: CDH1
Clonality: Monoclonal
Clone: 180224
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR