Anti-CD209B Rat Monoclonal Antibody [clone: 258829]

Supplier: R & D Systems
MAB1836
220019-391EA 577.02 USD
220019-391
Anti-CD209B Rat Monoclonal Antibody [clone: 258829]
Antibodies
Type: Primary
Antigen: CD209B
Clonality: Monoclonal
Clone: 258829
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR