Anti-IL27 Rat Monoclonal Antibody [clone: 355028]

Supplier: R & D Systems
MAB18343
102885-388EA 594.37 USD
102885-388
Anti-IL27 Rat Monoclonal Antibody [clone: 355028]
Antibodies
Type: Primary
Antigen: IL27
Clonality: Monoclonal
Clone: 355028
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR