Anti-IL27 Rat Monoclonal Antibody [clone: 355025]

Supplier: R & D Systems
MAB18342
102885-386EA 594.37 USD
102885-386
Anti-IL27 Rat Monoclonal Antibody [clone: 355025]
Antibodies
Type: Primary
Antigen: IL27
Clonality: Monoclonal
Clone: 355025
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR