Anti-IL6RA Rat Monoclonal Antibody [clone: 255820]

Supplier: R & D Systems
MAB18301
220019-387EA 600.15 USD
220019-387
Anti-IL6RA Rat Monoclonal Antibody [clone: 255820]
Antibodies
Type: Primary
Antigen: IL6RA
Clonality: Monoclonal
Clone: 255820
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR