Anti-CRP Rat Monoclonal Antibody [clone: 267917]

Supplier: R & D Systems
MAB1829
220019-384EA 772.25 USD
220019-384
Anti-CRP Rat Monoclonal Antibody [clone: 267917]
Antibodies
Type: Primary
Antigen: CRP
Clonality: Monoclonal
Clone: 267917
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR