Anti-TREM2 Mouse Monoclonal Antibody [clone: 263602]

Supplier: R & D Systems
MAB1828
220019-383EA 655.11 USD
220019-383
Anti-TREM2 Mouse Monoclonal Antibody [clone: 263602]
Antibodies
Type: Primary
Antigen: TREM2
Clonality: Monoclonal
Clone: 263602
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR