Anti-ERK1 + ERK2 Mouse Monoclonal Antibody [clone: 798508]

Supplier: R & D Systems
MAB1825
102885-382EA 600.15 USD
102885-382
Anti-ERK1 + ERK2 Mouse Monoclonal Antibody [clone: 798508]
Antibodies
Type: Primary
Antigen: ERK1 + ERK2
Clonality: Monoclonal
Clone: 798508
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR