Anti-CFD Mouse Monoclonal Antibody [clone: 255706]

Supplier: R & D Systems
MAB1824
220019-380EA 763.57 USD
220019-380
Anti-CFD Mouse Monoclonal Antibody [clone: 255706]
Antibodies
Type: Primary
Antigen: CFD
Clonality: Monoclonal
Clone: 255706
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR