Anti-S100 B Mouse Monoclonal Antibody [clone: 472806]

Supplier: R & D Systems
MAB1820
220019-379EA 594.37 USD
220019-379
Anti-S100 B Mouse Monoclonal Antibody [clone: 472806]
Antibodies
Simple Western

Type: Primary
Antigen: S100 B
Clonality: Monoclonal
Clone: 472806
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR