Anti-CCR5 Mouse Monoclonal Antibody [clone: 45531]

Supplier: R & D Systems
MAB182-100
102885-380EA 655.11 USD
102885-380
Anti-CCR5 Mouse Monoclonal Antibody [clone: 45531]
Antibodies
Type: Primary
Antigen: CCR5
Clonality: Monoclonal
Clone: 45531
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR