Anti-KL Rat Monoclonal Antibody [clone: 236214]

Supplier: R & D Systems
MAB1819
220019-377EA 772.25 USD
220019-377
Anti-KL Rat Monoclonal Antibody [clone: 236214]
Antibodies
Type: Primary
Antigen: KL
Clonality: Monoclonal
Clone: 236214
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR