Anti-DLL1 Mouse Monoclonal Antibody [clone: 251123]

Supplier: R & D Systems
MAB18181
220019-376EA 594.37 USD
220019-376
Anti-DLL1 Mouse Monoclonal Antibody [clone: 251123]
Antibodies
Type: Primary
Antigen: DLL1
Clonality: Monoclonal
Clone: 251123
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR