Anti-TNFRSF22 Rat Monoclonal Antibody [clone: 263310]

Supplier: R & D Systems
MAB1816
220019-373EA 577.02 USD
220019-373
Anti-TNFRSF22 Rat Monoclonal Antibody [clone: 263310]
Antibodies
Type: Primary
Antigen: TNFRSF22
Clonality: Monoclonal
Clone: 263310
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR