Anti-CHRDL1 Mouse Monoclonal Antibody [clone: 263737]

Supplier: R & D Systems
MAB1808
220019-371EA 561.11 USD
220019-371
Anti-CHRDL1 Mouse Monoclonal Antibody [clone: 263737]
Antibodies
Type: Primary
Antigen: CHRDL1
Clonality: Monoclonal
Clone: 263737
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR